Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix

Catalog No. p-4919371
Encompass_Preferred
Click to view available options
No. of Reactions:
250 Reactions
2500 Reactions

Includes

This master mix comes in a 250μL tube, enough for 25 reactions.

The Applied Biosystems AmpliTaq Gold Fast PCR Master Mix is a premix, hot start chemistry for end-point PCR analysis.

The Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix is a premix, hot start chemistry for end-point PCR analysis. Its optimized chemical design enables to shorten PCR running time from a traditional 2 hours to proximal 40 minutes. It generates high quality PCR products that give high quality sequencing results that customers expect from Applied Biosystems™. Optimized for use in sequencing applications, this kit provides specific, high-yield amplicons that can be easily and robustly sequenced. This premixed master mix works with your existing primers to provide sensitive, specific and reproducible results.

  • Fast Results—Speeds the time from sample to results. Amplifies 600bp length gene in approximately 45 minutes
  • Better Sequencing quality—Specific, high-yield amplicons suitable for sequencing applications that yield low peak-under-peak, longer continous read-length and higher QV-30 count
  • Easy to use—The 2X master mix allows for quick experiment setup
  • No primer annealing restrictions

Speed Time to Discovery

AmpliTaq Gold™ Fast PCR Master Mix, UP brings PCR reaction times down to as little as 45 minutes and delivers robust sequencing data on par with standard PCR reagents such as the classic AmpliTaq Gold™ PCR Master Mix. This new master mix has been designed for use with your existing primers and results in specific, high-yield amplicons for easy, high quality sequencing data. Cycle sequencing can now also be performed in as little as 50 minutes.

Quick Setup, High Sensitivity

This pre-mixed hotstart kit allows you to setup your experiment quickly without sacrificing performance. This kit provides reproducible, sensitive results, allowing you to detect a single copy of a gene in 10 ng of DNA.

Robust Fast Sequencing Results

Get the sequencing results you expect from Applied Biosystems™. The AmpliTaq Gold™ Fast PCR Master Mix, UP is optimized for your sequencing reactions. This kit yields specific high-yield amplicons that provide you with clean sequencing reads including low peak-under-peak, longer read-lengths and superior Q30.

Note: See user's manual or package insert for limited label license, and trademark information.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • AmpliTaq Gold Fast PCR Master Mix, 2 x 1.25 mL tubes

Store at 2°C to 8°C.
Format Tube
GC-Rich PCR Performance High
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Fast
Exonuclease Activity 5' - 3'
Product Type Fast PCR Master Mix
Quantity 250 reactions
Shipping Condition Wet Ice
For Use With (Application) Cloning, Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 250 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Size (Final Product) 1.5 kb or less
Starting Material DNA
Volume 2.5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.