Learn More
Applied Biosystems™ SYBR™ Select Master Mix for CFX
A cost-effective solution for real-time PCR applications.
$133.00 - $4720.00
Specifications
Concentration | 2X |
---|---|
Detection Method | SYBR |
Form | Liquid |
Format | Vial |
GC-Rich PCR Performance | High |
Includes: 2X mix contains SYBR GreenER dye, AmpliTaq DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, and optimized buffer components.
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
44-729-37
![]() |
Applied Biosystems™
4472937 |
1 x 1 mL |
Each for $133.00
|
|
|||||
44-729-42
![]() |
Applied Biosystems™
4472942 |
1 x 5 mL |
Each for $556.00
|
|
|||||
44-729-52
![]() |
Applied Biosystems™
4472952 |
2 x 5 mL |
Each for $1,068.00
|
|
|||||
44-729-54
![]() |
Applied Biosystems™
4472954 |
10 x 5 mL |
Each for $4,720.00
|
|
|||||
44-729-53
![]() |
Applied Biosystems™
4472953 |
5 x 5 mL |
Each for $2,470.00
|
|
|||||
Description
- Specific - minimize primer-dimer and non-specific amplification
- Reproducible and sensitive - consistent amplification across a wide dynamic range
- Bright - contains SYBR™ GreenER™ dye for maximum brightness
- Carry-over contamination control - contains heat-labile UDG
- For use with the Bio-Rad CFX96™ Touch or CFX384™ Touch systems
- Standard and fast cycling parameters
Formulated for Maximum Specificity:
- SYBR™ Select Master Mix for CFX contains all the components needed for your real-time PCR reactions, except the template and primers, in a convenient 2X concentration premix
- The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity
Obtain Reproducible Results Across a Wide Dynamic Range:
- SYBR™ Select Master Mix for CFX is specially formulated to provide robust results from 100ng to 0.1pg cDNA per reaction and can reliably detect a single copy of genomic DNA.
- Contains SYBR™ GreenER™ Dye and Heat-labile UDG:
- SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye, resulting in brighter signals
- Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control
Order Info
Shipping Conditions: Wet Ice
Specifications
2X | |
Liquid | |
High | |
AmpliTaq DNA Polymerase | |
AB 7900HT, Standard Mode, AB StepOne™, Fast Mode, AB 7500, Standard Mode, AB 7500, Fast Mode, Bio-Rad CFX96 Touch™, AB StepOnePlus™, Fast Mode, AB 7900HT, Fast Mode, AB StepOne™, Standard Mode, Bio-Rad CFX Connect™, AB 7900 HT 384-Well System, Bio-Rad CFX384 Touch™, AB StepOnePlus™, Standard Mode | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
DNA (Genomic), cDNA |
SYBR | |
Vial | |
qPCR | |
Fast or Standard | |
SYBR GreenER | |
SYBR™ | |
UP (Ultra Pure) | |
Built-In Hot Start |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.