Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APR3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179969
Description
44289 Polyclonal specifically detects 44289 in Human samples. It is validated for Western Blot.Specifications
APR3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
apoptosis related protein 3, apoptosis related protein APR-3, apoptosis-related protein 3, APR3, APR-3, APR--3, chromosome 2 open reading frame 28, HSPC013, p18PRO240 | |
Rabbit | |
18 kDa | |
100 μL | |
Apoptosis | |
51374 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_057169 | |
ATRAID | |
Synthetic peptide directed towards the C terminal of human C2orf28. Peptide sequence VCADGFHGYKCMRQGSFSLLMFFGILGATTLSVSILLWATQRRKAKTS. | |
Affinity purified | |
RUO | |
Primary | |
Porcine: 87%; Rat: 87%; Mouse: 87%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction