Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APXL Antibody, Novus Biologicals™
SDP

Catalog No. NBP215437 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215437 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215437 Supplier Novus Biologicals Supplier No. NBP215437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APXL Polyclonal antibody specifically detects APXL in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen APXL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q13796
Gene Alias apical protein, Xenopus laevis-like, apical protein-like (Xenopus laevis), Apical-like protein, APX homolog of Xenopus, APXLHSAPXL, DKFZp781J074, FLJ39277, Protein APXL, protein Shroom2, shroom family member 2
Gene Symbols SHROOM2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human APXL. The exact sequence is proprietary.
Molecular Weight of Antigen 176 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 357
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse (85%) and Rat (85%).
Target Species Human, Mouse, Monkey
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.