Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ APY 29
GREENER_CHOICE

Catalog No. 486510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-651-0 10 mg
48-655-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-651-0 Supplier Tocris Bioscience™ Supplier No. 4865/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits IRE1α autophosphorylation; activates IRE1α endoribonuclease activity

Allosteric modulator of IRE1α. Inhibits IRE1α autophosphorylation (IC50 = 280 nM) and activates IRE1α ribonuclease activity.

Chemical Identifiers

CAS 1216665-49-4
Molecular Formula C17H16N8

Specifications

Quantity 10 mg
Formula Weight 332.36
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 332.371
Chemical Name or Material APY 29
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.