Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 1/AQP1 Antibody (OTI2D10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270199V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270199V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270199V Supplier Novus Biologicals Supplier No. NBP270199V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aquaporin 1/AQP1 Monoclonal specifically detects Aquaporin 1/AQP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Aquaporin 1/AQP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2D10
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324
Gene Symbols AQP1
Host Species Mouse
Immunogen Full length recombinant protein of human Aquaporin 1/AQP1 (NP_932766) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 358.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.