Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP170378 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP170378 0.1 mL
NB018704 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP170378 Supplier Novus Biologicals Supplier No. NBP170378
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

Aquaporin-2 Polyclonal specifically detects Aquaporin-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Aquaporin-2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:300-1:2000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P56402, P34080
Gene Alias ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD
Gene Symbols AQP2
Host Species Goat
Immunogen A synthetic peptide from human Aquaporin-2 conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 359
Test Specificity Human, rat, mouse. Other species not yet tested.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.