Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-2 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody has been used in 3 publications

$206.50 - $489.50

Specifications

Antigen Aquaporin-2
Concentration LYOPH
Classification Polyclonal
Conjugate Unconjugated
Form Antisera
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018704
SDP
View Documents
Novus Biologicals
NBP1703780.025ML
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP170378
SDP
View Documents
Novus Biologicals
NBP170378
0.1 mL
Each for $489.50
Only null left
Add to Cart
 
Description

Description

Aquaporin-2 Polyclonal specifically detects Aquaporin-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Aquaporin-2
Polyclonal
Antisera
RUO
Rat, Human, Mouse
ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD
AQP2
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
LYOPH
Unconjugated
Goat
Plasma Membrane Markers, Signal Transduction
P56402, P34080
359
IgG
Unpurified
Human, rat, mouse. Other species not yet tested.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.