Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 3 Rabbit anti-Bovine, Human, Mouse, Ovine, Rat, Polyclonal, FabGennix

Catalog No. 502206105
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6105 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6105 Supplier Fabgennix Inc Supplier No. AQP3301AP
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aquaporin 3 Polyclonal antibody specifically detects Aquaporin 3 in Bovine, Human, Mouse, Ovine, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Aquaporin 3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene AQP3
Gene Accession No. P47862, Q08DE6, Q8R2N1, Q92482
Gene Alias 31.4 kDa water channel protein; AQP 3; AQP-2; AQP3; AQP-3; Aquaglyceroporin-3; aquaporin 3; aquaporin 3 (GIL blood group); aquaporin 3 (Gill blood group); Aquaporin3; aquaporin-3; GIL; Gill blood group
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 200-250 on rat Aquaporin 3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11828, 360, 443047, 65133, 780866
Target Species Bovine, Human, Mouse, Ovine, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.