Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$416.50 - $684.50
Specifications
Antigen | Aquaporin-4 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Aquaporin-4 Polyclonal specifically detects Aquaporin-4 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Aquaporin-4 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat, Bovine | |
P55087 | |
361 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CPDVEFKRRFKEAFSKAAQQTKGSYMEVEDNRSQVETDDLILKPGVVHVIDVDRGEEKKGKDQSGEVLSSV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
35 kDa |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
aquaporin 4, MGC22454 | |
AQP4 | |
IgG | |
Affinity Purified | |
Specificity of human Aquaporin-4 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title