Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-5 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP267247A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP267247A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP267247A94 Supplier Novus Biologicals Supplier No. NBP267247AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aquaporin-5 Polyclonal specifically detects Aquaporin-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Aquaporin-5
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AQP-5, aquaporin 5, aquaporin-5
Gene Symbols AQP5
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of the human Aquaporin-5 protein (between amino acids 170-250). [UniProt P55064]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 362.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.