Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARA54 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215438 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215438 Supplier Novus Biologicals Supplier No. NBP215438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARA54 Polyclonal specifically detects ARA54 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ARA54
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UBS8
Gene Alias Androgen receptor-associated protein 54, ARA54androgen receptor associated protein 54, EC 6.3.2.-, HFB30E3 ubiquitin-protein ligase RNF14, ring finger protein 14FLJ26004, TRIAD2, Triad2 protein
Gene Symbols RNF14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNF14. The exact sequence is proprietary.
Molecular Weight of Antigen 54 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 9604
Test Specificity Human. Expected to cross react based on sequence identity: Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.