Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARAP1 Antibody (ARAP1-2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB359802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359802 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359802 Supplier Novus Biologicals Supplier No. NB11068801MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARAP1 Monoclonal antibody specifically detects ARAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen ARAP1
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone ARAP1-2
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2
Host Species Mouse
Immunogen His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 116985
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.