Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARAP3 Antibody, Novus Biologicals™
SDP

Catalog No. NB432559 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432559 25 μL
NBP184541 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB432559 Supplier Novus Biologicals Supplier No. NBP18454125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARAP3 Polyclonal specifically detects ARAP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ARAP3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8WWN8
Gene Alias Arf and Rho GAP adapter protein 3, arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 3, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3, ARF-GAP, RHO-GAP, ankyrin repeat and plekstrin homology domains-containingprotein 3, centaurin, delta 3, centaurin-delta-3, CENTD3, Cnt-d3, DRAG1, FLJ21065, phosphoinositide binding protein, PtdIns(34,5)P3-binding protein
Gene Symbols ARAP3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:MPNSIYFGLDSRGRAQAAQDKAPDSSQISAPTPALRPTTGTVHIMDPGCLYYGVQPVGTPGAPDRRESRGVCQGRAEHRLSRQDLEAREDAGYASLELPGDSTLLSPTLETEETSDDLISPYASFSF
Molecular Weight of Antigen 170 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64411
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.