Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARAP3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP259087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259087 Supplier Novus Biologicals Supplier No. NBP259087

Rabbit Polyclonal Antibody

ARAP3 Polyclonal specifically detects ARAP3 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen ARAP3
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias Arf and Rho GAP adapter protein 3, arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 3, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3, ARF-GAP, RHO-GAP, ankyrin repeat and plekstrin homology domains-containingprotein 3, centaurin, delta 3, centaurin-delta-3, CENTD3, Cnt-d3, DRAG1, FLJ21065, phosphoinositide binding protein, PtdIns(34,5)P3-binding protein
Gene Symbols ARAP3
Host Species Rabbit
Immunogen Maps to a region between 1200 and 1250
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64411
Target Species Human
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.