Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)

Catalog No. p-7075635
Encompass_Preferred
Click to view available options
No. of Tests:
25
100

ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)

ArC™ Amine Reactive Compensation Bead Kit is a bead-based compensation tool specifically optimized for use with LIVE/DEAD™ Fixable dead cell stain kits.

  • Ready-to-use control
  • Eliminates the hassle of heat-treating cells as a control, saving precious sample
  • Enables accurate and consistent results

    View a selection guide for all fixable viability dyes for flow cytometry.

    Ready-to-use control
    Unlike other methods that require sample preparation to make compensation controls using cellular material, the ArC™ Amine Reactive Compensation Bead Kit is ready to use out of the dropper bottle. The kit includes two types of specially modified polystyrene microspheres that have been optimized to react with the amine-reactive dye in the LIVE/DEAD™ Fixable dead cell stain kits. The ArC™ reactive beads (Component A) bind any of the amine-reactive dyes and provide a positive signal; the ArC™ negative beads (Component B) have no reactivity and provide a negative compensation control. This ready-to-use product preserves the original sample material for experiments; precious sample is not used for the creation of compensation controls.

    Accurate and consistent results
    Unlike biological controls, which can have variable expression levels depending upon disease or development state, the Arc™ beads provide a bright, consistent signal (see figure below). Consistency in the staining pattern of compensation controls is important for obtaining the right compensation value. If the signal varies with the control material used, the compensation value can be affected, leading to erroneous results.

    How it works
    One drop of beads is added to a test tube containing LIVE/DEAD™ Fixable dead cell stain. After incubation with amine-reactive dye, the beads are washed in staining buffer, a drop of negative control beads is added (if required), and the beads are resuspended and analyzed by flow cytometry. The two components provide a distinct positive bead that is at least 50-fold brighter than the negative beads, which can be used to set compensation.

    Other compensation products
    Besides the ArC™ Amine Reactive Compensation Beads, Life Technologies also offers the AbC™ Anti-Mouse Bead Kit for capture of mouse anti-human antibodies and AbC™ Anti-Rat/Hamster Bead Kit for capture of rat anti-mouse or hamster anti-mouse antibodies.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Detection Method Fluorescence
    Product Line CompenFlow™, Molecular Probes™
    Format Tube
    Excitation Wavelength Range any lasers (UV to 633/635)
    Content And Storage Contains 1 vial ArC™ amine reactive beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Diameter (Metric) 6 μm
    For Use With (Equipment) Flow Cytometer
    No. of Tests 100
    Quantity 1 Kit
    Shipping Condition Room Temperature
    Stain Configuration Surface
    Product Type Compensation Beads
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.