Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Arctigenin

Click to view available options
Quantity:
10 mg
50 mg
Description
Antioxidant, anti-inflammatory, antiproliferative and antiviral agent. Inhibits LPS-induced iNOS expression via inhibition of IκBα phosphorylation and p65 nuclear translocation (IC50 = 10 nM). Inhibits HIV-1 replication in vitro. Also potently inhibits MKK1 (MEK1) (IC50 = 0.5 nM) and provides neuroprotection by binding to kainate receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 372.42 |
| Purity | >98% |
| Molecular Weight (g/mol) | 372.417 |
| Chemical Name or Material | Arctigenin |
Safety and Handling
| Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction