Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARD1A Monoclonal Antibody (10)
GREENER_CHOICE

Catalog No. PIMA529432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529432 Supplier Invitrogen™ Supplier No. MA529432
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ARD1.

N-alpha-acetylation is one of the most common protein modifications that occurs during protein synthesis and involves the transfer of an acetyl group from acetyl-coenzyme A to the protein alpha-amino group. ARD1A, together with NATH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARD1A
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NAA10
Gene Accession No. P41227
Gene Alias 2310039H09Rik; alpha-N-acetyltransferase 1A; ARD1; ARD1 homolog A, N-acetyltransferase; Ard1a; ARD1P; DXS707; hARD1; MCOPS1; N(alpha)-acetyltransferase 10, NatA catalytic subunit; N(alpha)-acetyltransferase 10, NatA catalytic subunitNalpha acetyltransferase 10; Naa10; N-acetyltransferase ARD1 homolog; N-acetyltransferase ARD1, human homolog of; Nalpha acetyltransferase 10; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; natA catalytic subunit Naa10; NATD; N-terminal acetyltransferase complex ARD1 subunit homolog A; OGDNS; RGD1565315; Te2
Gene Symbols NAA10
Host Species Mouse
Immunogen Recombinant Human ARD1 protein (Met1-Ser235).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.