Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1,2,3,4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10055421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10055421 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10055421 Supplier Novus Biologicals Supplier No. NB10055421
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ARF1,2,3,4 Polyclonal specifically detects ARF1,2,3,4 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Peptide ELISA, Western Blot.

Specifications

Antigen ARF1,2,3,4
Applications Flow Cytometry, Peptide ELISA, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 μg/mL, Flow Cytometry 10 μg/mL, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001019397.1
Gene Alias ADP-ribosylation factor 1
Gene Symbols ARF1
Host Species Goat
Immunogen Peptide with sequence C-EGLDWLSNQLRNQK, from the C Terminus of the protein sequence according to NP_001019397.1; NP_001649.1; NP_001019398.1; NP_001019399.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375
Test Specificity This antibody is expected to recognize all four reported isoforms.
Target Species Canine, Human, Rat, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.