Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Antibody (ARF1/2117), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB386706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386706 Supplier Novus Biologicals Supplier No. NBP308473AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARF1 Monoclonal antibody specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ARF1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ARF1/2117
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factor 1
Host Species Mouse
Immunogen Recombinant full-length human ARF1 protein (Uniprot: P84077)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 375
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.