Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Antibody (ARFS 1A9/5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N102635A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102635A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102635A594 Supplier Novus Biologicals Supplier No. NB1002635AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARF1 Monoclonal specifically detects ARF1 in Human samples. It is validated for ELISA, Immunofluorescence, Western Blot.

Specifications

Antigen ARF1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone ARFS 1A9/5
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ADP-ribosylation factor 1
Gene Symbols ARF1
Host Species Mouse
Immunogen C-terminal Human ARF1 peptide SNQLRNQ
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 375
Test Specificity ARFS 1A9/5 is Class 1 ARF specific.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.