Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Antibody (ARFS 3F1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB185530646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185530646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB185530646 Supplier Novus Biologicals Supplier No. NB11085530JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARF1 Monoclonal antibody specifically detects ARF1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen ARF1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone ARFS 3F1
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ADP-ribosylation factor 1
Gene Symbols ARF1
Host Species Mouse
Immunogen C-terminal Human ARF1 peptide SNQLRNQ
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 375
Test Specificity ARFS 3F1 is Class 1 ARF specific.
Target Species Human, Canine, Chicken, Guinea Pig, Mouse, Pig, Primate, Rabbit, Rat, Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.