Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Mouse anti-Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Porcine, Primate, Rabbit, Rat, Sheep, Clone-ARFS 3F1, Novus Biologicals™
SDP

Catalog No. NB110855301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB110855301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB110855301 Supplier Novus Biologicals Supplier No. NB110855300.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARF1 Monoclonal specifically detects ARF1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ARF1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone ARFS 3F1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P84077
Gene Alias ADP-ribosylation factor 1
Gene Symbols ARF1
Host Species Mouse
Immunogen C-terminal Human ARF1 peptide SNQLRNQ
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375
Test Specificity ARFS 3F1 is Class 1 ARF specific.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.