Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179614
Description
ARF4 Polyclonal specifically detects ARF4 in Mouse samples. It is validated for Western Blot.Specifications
ARF4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ADP-ribosylation factor 4, ARF2ADP-ribosylation factor 2 | |
Rabbit | |
20 kDa | |
100 μL | |
Primary | |
This antibody is predicted to react to human ARF1,3,4,5 (100% homology) and 6 (93%). | |
Mouse, Rat, Pig, Equine, Yeast, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_031505 | |
ARF4 | |
Synthetic peptide directed towards the N terminal of human Arf4The immunogen for this antibody is Arf4. Peptide sequence VGLDAAGKTTILYKLKLGEIVTTIPTIGFNVETVEYKNICFTVWDVGGQD. | |
Affinity purified | |
RUO | |
378 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction