Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arfaptin-1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Arfaptin-1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8367
![]() |
Bethyl Laboratories, Inc
A304676AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Arfaptin 1 is an ARF binding protein that inhibits PLD activation, vesicular trafficking and secretion. In the present report, we show that arfaptin 1 interacts with 'high speed' membranes independently of ARF. However, addition of myristoylated ARF3 (myrARF3) increases the association of arfaptin 1 with the membranes, suggesting that arfaptin 1 and ARF form a complex on the Golgi. Utilizing several deletion mutants of arfaptin 1 it is shown that the association of arfaptin 1 with myrARF3 is mediated via two binding sites on arfaptin 1. These two domains are needed for arfaptin 1 inhibition of PLD activation by myrARF3 in vitro.Specifications
Arfaptin-1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
HSU52521, ADP-ribosylation factor-interacting protein 1, arfaptin 1, arfaptin-1 | |
Between 1 and 50 | |
Primary | |
4° C | |
ARFIP1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P53367 | |
27236, 99889 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title