Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARFGAP3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP118921 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118921 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP118921 Supplier Novus Biologicals Supplier No. NBP118921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARFGAP3 Polyclonal specifically detects ARFGAP3 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen ARFGAP3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9NP61
Gene Alias ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase activating protein 3, ADP-ribosylation factor GTPase-activating protein 3, ARF GAP 3, ARFGAP1, FLJ45618
Gene Symbols ARFGAP3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 466 and 516 of human ADP-ribosylation factor GTPase activating protein 3 using the numbering given in entry NP_055385.2 (GeneID 26286).
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26286
Test Specificity Human. Expected cross-reactivity based on sequence identity (not validated): Guinea pig (100%), Orangutan (100%), Rhesus Monkey (100%), Gorilla (100%), Chimpanzee (100%), Nake mole rat (100%), and Northern white-cheeked gibbon (100%).
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.