Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARG2 Antibody (7F8), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB160592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160592 Supplier Novus Biologicals Supplier No. NBP259406DL594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARG2 Monoclonal antibody specifically detects ARG2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen ARG2
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 7F8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias arginase 2, arginase, type II, arginase-2, mitochondrial, EC 3.5.3, EC 3.5.3.1, kidney arginase, Kidney-type arginase, L-arginine amidinohydrolase, L-arginine ureahydrolase, nonhepatic arginase, Non-hepatic arginase, Type II arginase
Host Species Mouse
Immunogen Recombinant human ARG2 (23-354aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 384
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.