Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Argatroban

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.
This compound is a mixture of 2 diastereomers (approximate ratio 65/35).
Specifications
Specifications
| Chemical Name or Material | Argatroban |
| CAS | 74863-84-6 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Molecular Formula | C23H36N6O5S |
| Formula Weight | 508.63 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction