Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 11 publications
$254.50 - $584.50
Specifications
Antigen | Arginase 1/ARG1/liver Arginase |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10059740
![]() |
Novus Biologicals
NB10059740 |
0.1 mg |
Each for $584.50
|
|
|||||
NB407643
![]() |
Novus Biologicals
NB1005974025UG |
25 μg |
Each for $254.50
|
|
|||||
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA.Specifications
Arginase 1/ARG1/liver Arginase | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
ARG1 | |
Primary | |
37 kDa |
0.5 mg/mL | |
Polyclonal | |
Goat | |
Human, Mouse, Rat | |
P07824 | |
383 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title