Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginase 1/ARG1/liver Arginase Antibody (658922), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347219 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347219 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347219 Supplier Novus Biologicals Supplier No. FAB58681MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Arginase 1/ARG1/liver Arginase Monoclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Flow Cytometry, CyTOF-reported

Specifications

Antigen Arginase 1/ARG1/liver Arginase
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 658922
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase
Host Species Mouse
Immunogen E. coli-derived recombinant human Arginase 1/ARG1, Met1-Lys322, Accession # P05089
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 383
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.