Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginase 1/ARG1/liver Arginase Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB132731PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132731PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132731PCP Supplier Novus Biologicals Supplier No. NBP132731PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockdown Validated.

Specifications

Antigen Arginase 1/ARG1/liver Arginase
Applications Flow Cytometry, KnockDown
Classification Polyclonal
Conjugate PerCP
Dilution Flow Cytometry, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Gene Symbols ARG1
Host Species Rabbit
Immunogen Arginase 1/ARG1/liver Arginase Antibody made from full length human Arginase 1 Recombinant protein.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 383.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.