Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Arginase 2 Monoclonal Antibody (D3)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | Arginase 2 |
---|---|
Clone | D3 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541557
|
Invitrogen™
MA541557 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543949
|
Invitrogen™
MA543949 |
20 μL |
Each for $243.00
|
|
|||||
Description
Arginase 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Arginase catalyzes the hydrolysis of arginine to ornithine and urea. At least two isoforms of mammalian arginase exists (types I and II) which differ in their tissue distribution, subcellular localization, immunologic crossreactivity and physiologic function. The type II isoform encoded by this gene, is located in the mitochondria and expressed in extra-hepatic tissues, especially kidney. The physiologic role of this isoform is poorly understood; it is thought to play a role in nitric oxide and polyamine metabolism. Transcript variants of the type II gene resulting from the use of alternative polyadenylation sites have been described.Specifications
Arginase 2 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P78540 | |
384 | |
Recombinant protein Arginase II. The antigen corresponds to amino acid range 23-354 of the target protein. | |
Antibody |
D3 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
ARG2 | |
AII; AII type II arginase; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; nonhepatic arginase; non-hepatic arginase; type II arginase | |
ARG2 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title