Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argininosuccinate Synthase Antibody (1G11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB161427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161427 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161427 Supplier Novus Biologicals Supplier No. NBP259443AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Argininosuccinate Synthase Monoclonal antibody specifically detects Argininosuccinate Synthase in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Argininosuccinate Synthase
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1G11
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5
Host Species Mouse
Immunogen Recombinant human Argininosuccinate Synthase (1-412aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 445
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.