Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argininosuccinate Synthase Antibody (OTI1B10), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP270217UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270217UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270217UV Supplier Novus Biologicals Supplier No. NBP270217UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Argininosuccinate Synthase Monoclonal specifically detects Argininosuccinate Synthase in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Argininosuccinate Synthase
Applications Western Blot
Classification Monoclonal
Clone OTI1B10
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5
Gene Symbols ASS1
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues near C-terminus of human Argininosuccinate Synthase (NP_000041).
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 445.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.