Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARH Antibody (OTI7E11), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP271790C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271790C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271790C Supplier Novus Biologicals Supplier No. NBP271790C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARH Monoclonal specifically detects ARH in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ARH
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI7E11
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705
Gene Symbols LDLRAP1
Host Species Mouse
Immunogen Full length recombinant protein of human ARH (NP_056442) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 26119.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.