Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224707SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224707SS 0.025 mg
NBP224707 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224707SS Supplier Novus Biologicals Supplier No. NBP224707SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARHGAP1 Polyclonal specifically detects ARHGAP1 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.

Specifications

Antigen ARHGAP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Accession No. Q07960
Gene Alias CDC42 GTPase-activating protein, CDC42GAPp50-RhoGAP, GTPase-activating protein rhoOGAP, p50rhoGAP, Rho GTPase activating protein 1, rho GTPase-activating protein 1, RHOGAP, RHOGAP1Cdc42GAP, Rho-related small GTPase protein activator, Rho-type GTPase-activating protein 1
Gene Symbols ARHGAP1
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human RhoGAP1 was used as the immunogen
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 392
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat (each having multiple isoforms), 94% homologous in bovine, 88% homologous in chicken, 76% homologous in Xenopus, and 70% homologous in zebrafish.
Target Species Human, Mouse, Rat, Bovine, Canine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.