Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP17 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB042728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042728 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042728 Supplier Novus Biologicals Supplier No. NBP297377JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARHGAP17 Polyclonal specifically detects ARHGAP17 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ARHGAP17
Applications Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp564A1363, FLJ10308, FLJ13219, FLJ37567, FLJ43368, MGC87805, MST110, MSTP038, MSTP066, MSTP110, NADRIN, neuron-associated developmentally regulated protein, PP367, PP4534, Rho GTPase activating protein 17, rho GTPase-activating protein 17, RhoGAP interacting with CIP4 homologs 1, RhoGAP interacting with CIP4 homologs protein 1, Rho-type GTPase-activating protein 17, RICH-1, RICH1B, RICH1MST066, WBP15
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ARHGAP17 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55114
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.