Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP18 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342731 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342731 Supplier Novus Biologicals Supplier No. NBP281704MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARHGAP18 Polyclonal antibody specifically detects ARHGAP18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ARHGAP18
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bA307O14.2, MacGAP, MGC126757, Rho GTPase activating protein 18, rho-type GTPase-activating protein 18
Host Species Rabbit
Immunogen ARHGAP18 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ARHGAP18. The immunogen is located within the last 50 amino acids of ARHGAP18. Amino Acid Squence: DTYMKDLYQLNPNAE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 93663
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.