Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP21 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB146537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB146537 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB146537 Supplier Novus Biologicals Supplier No. NBP306052AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARHGAP21 Polyclonal specifically detects ARHGAP21 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ARHGAP21
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp761L0424, FLJ33323, FLJ90108, KIAA1424, Rho GTPase activating protein 21, rho GTPase-activating protein 10, rho GTPase-activating protein 21, Rho-GTPase activating protein 10, rho-type GTPase-activating protein 21
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ARHGAP21 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57584
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.