Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP22, p Ser397 Antibody, Novus Biologicals™
SDP

Catalog No. NBP144073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144073 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144073 Supplier Novus Biologicals Supplier No. NBP144073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARHGAP22 (p Ser397) Polyclonal specifically detects ARHGAP22 (p Ser397) in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen ARHGAP22 (p Ser397)
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with 1 mg/ml Bovine Serum Albumin (BSA) with No Preservative
Gene Accession No. NP_722495, Q8BL80
Gene Alias Rho GTPase activating protein 22, rho GTPase-activating protein 22, RHOGAP2, Rho-type GTPase-activating protein 22
Gene Symbols ARHGAP22
Host Species Rabbit
Immunogen ARHGAP22 [p Ser397] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic phospho-peptide corresponding to the region surrounding mouse pS397 region of ARHGAP22. (Uniprot: Q8BL80)
Molecular Weight of Antigen 77.78 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 58504
Test Specificity This antibody is specific for phosphorylated ARHGAP22 at Serine 397. It also recognizes the S22->A mutation but not the S397->A mutation. A BLAST analysis was used to suggest cross-reactivity with Mouse, Rat and Human based on 100% sequence homology. Cross-reactivity with ARHGAP22 pS397 from other sources has not been determined.
Reconstitution Reconstitute in 185μL of water if being used immediately.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.