Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP25 Antibody (OTI2D5), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N271508PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271508PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271508PEC7 Supplier Novus Biologicals Supplier No. NBP271508PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARHGAP25 Monoclonal specifically detects ARHGAP25 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen ARHGAP25
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2D5
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias KAIA0053, KIAA0053, Rho GTPase activating protein 25, rho GTPase-activating protein 25, Rho-type GTPase-activating protein 25
Gene Symbols ARHGAP25
Host Species Mouse
Immunogen Full length recombinant protein of human ARHGAP25 (NP_055697) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9938.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.