Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB027535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB027535 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB027535 Supplier Novus Biologicals Supplier No. NBP29283420UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

ARHGEF3 Polyclonal antibody specifically detects ARHGEF3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ARHGEF3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZP434F2429, Exchange factor found in platelets and leukemic and neuronal tissues, FLJ98126, MGC118905, Rho guanine nucleotide exchange factor (GEF) 3, rho guanine nucleotide exchange factor 3,59.8 kDA protein, RhoGEF protein, XPLNXPLN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 287-526 of human ARHGEF3 (NP_062455.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50650
Target Species Human, Mouse
Content And Storage Store at -20°:C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.