Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF4 Antibody (ARHGEF-08), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB002671 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002671 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002671 Supplier Novus Biologicals Supplier No. NBP262217R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARHGEF4 Monoclonal specifically detects ARHGEF4 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen ARHGEF4
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone ARHGEF-08
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias APC-stimulated guanine nucleotide exchange factor 1, Asef, ASEF1, ASEFAPC-stimulated guanine nucleotide exchange factor, GEF4, KIAA1112, Rho guanine nucleotide exchange factor (GEF) 4, rho guanine nucleotide exchange factor 4, STM6
Gene Symbols ARHGEF4
Host Species Mouse
Immunogen Recombinant fragment of human ARHGEF4 (amino acids 143-271)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50649
Test Specificity The antibody MEM-174 recognizes CD11b antigen (Mac-1 alpha), a 165-170 kDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.