Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF4 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001316 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001316 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001316 Supplier Novus Biologicals Supplier No. NB1001316
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

ARHGEF4 Polyclonal specifically detects ARHGEF4 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen ARHGEF4
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 4 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9NR80
Gene Alias APC-stimulated guanine nucleotide exchange factor 1, Asef, ASEF1, ASEFAPC-stimulated guanine nucleotide exchange factor, GEF4, KIAA1112, Rho guanine nucleotide exchange factor (GEF) 4, rho guanine nucleotide exchange factor 4, STM6
Gene Symbols ARHGEF4
Host Species Goat
Immunogen Peptide with sequence PWEEPAGEKPSCS corresponding to N-Terminus according to NP_056135.2, NP_127462.1.
Molecular Weight of Antigen 79 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50649
Test Specificity This antibody is expected to recognize both reported isoforms (NP_056135.2; NP_127462.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.