Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID1A Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291854 Supplier Novus Biologicals Supplier No. NB10055334PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARID1A Polyclonal specifically detects ARID1A in Human, Mouse, Zebrafish samples. It is validated for ChIP Assay, Immunoprecipitation (Negative), Western Blot.

Specifications

Antigen ARID1A
Applications ChIP Assay, Immunoprecipitation (Negative), Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation (Negative), Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias ARID domain-containing protein 1A, AT rich interactive domain 1A (SWI- like), AT rich interactive domain 1A (SWI-like), AT-rich interactive domain-containing protein 1A, B120SWI-like protein, BAF250a, BAF250SWI/SNF complex protein p270, BM029, brain protein 120, BRG1-associated factor 250, BRG1-associated factor 250a, C10rf4, C1orf4SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatinsubfamily F member 1, chromatin remodeling factor p250, hELD, hOSA1, matrix associated, actin dependent regulator of chromatin, Osa homolog 1, OSA1, OSA1 nuclear protein, P270, SMARCF1, subfamily f, member 1
Host Species Rabbit
Immunogen Maps to a region between residue 750 and 800 of human AT rich interactive domain 1A (BRG1-associated factor 250kD) using the numbering given in entry NP_006006.3 (GeneID 8289).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8289
Target Species Human, Mouse, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.