Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID1B Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB049065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB049065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB049065 Supplier Novus Biologicals Supplier No. NBP297890R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARID1B Polyclonal specifically detects ARID1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ARID1B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 6A3-5, ARID domain-containing protein 1B, AT rich interactive domain 1B (SWI1-like), BAF250B, BRG1-associated factor 250b, BRG1-binding protein ELD/OSA1, BRG1-binding protein hELD/OSA1, BRIGHT, DAN15P250R, ELD (eyelid)/OSA protein, ELD/OSA1, hOsa2, JPUT1, KDM5B, KIAA1235AT-rich interactive domain-containing protein 1B, Osa homolog 2, OSA2, p250R, PLU1, RBBP2H1A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ARID1B fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57492
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.