Learn More
Description
Ensure accurate, reproducible results in Western Blot
Specifications
Specifications
| Antigen | ARID2 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay |
| Classification | Monoclonal |
| Clone | 7311 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Knockout Validated |
| Formulation | PBS with No Preservative |
| Gene Alias | ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200 |
| Gene Symbols | ARID2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
