Learn More
Description
Specifications
Specifications
| Antigen | ARID2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200 |
| Gene Symbols | ARID2 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 1785 and 1835 of human AT rich interactive domain 2 using the numbering given in entry NP_689854.2 (GeneID 196528). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
