Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID3B Antibody (008), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB308363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308363 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308363 Supplier Novus Biologicals Supplier No. NBP289222APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ARID3B Monoclonal antibody specifically detects ARID3B in Human samples. It is validated for Western Blot

Specifications

Antigen ARID3B
Applications Western Blot
Classification Monoclonal
Clone 008
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias ARID domain-containing protein 3B, AT rich interactive domain 3B (BRIGHT- like), AT rich interactive domain 3B (BRIGHT-like), AT-rich interactive domain-containing protein 3B, BDP, bright and dead ringer protein, DRIL2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human ARID3B.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10620
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.