Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID3C Antibody, Novus Biologicals™
SDP

Catalog No. NBP120885 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120885 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120885 Supplier Novus Biologicals Supplier No. NBP120885
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ARID3C Polyclonal specifically detects ARID3C in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen ARID3C
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001017363.1
Gene Alias ARID domain-containing protein 3C, AT rich interactive domain 3C (BRIGHT- like), AT rich interactive domain 3C (BRIGHT-like), AT-rich interactive domain-containing protein 3C
Gene Symbols ARID3C
Host Species Goat
Immunogen Peptide with sequence C-REKLAPEEPPEKR corresponding to internal region according to NP_001017363.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138715
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.